missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LBP Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB6635-SP
This item is not returnable.
View return policy
Description
LBP Monoclonal specifically detects LBP in Mouse samples. It is validated for Neutralization.
Specifications
| LBP | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| lipopolysaccharide binding protein, lipopolysaccharide-binding protein, LPS-binding protein, MGC22233 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3929 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization | |
| 749405 | |
| Neutralization 1-5 ug/mL | |
| Q61805 | |
| LBP | |
| Mouse myeloma cell line NS0-derived recombinant mouse LBP Gly25-Val481 (Ser102Arg, Tyr284His) Accession # Q61805 | |
| 25 μg | |
| Primary | |
| Detects mouse LBP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LBP is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction