missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Laminin alpha 1/beta 1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4656-SP
This item is not returnable.
View return policy
Description
Laminin alpha 1 Monoclonal specifically detects Laminin alpha 1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Laminin alpha 1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Lama1 | |
| Purified fragment of chymotrypsin-digested mouse EHS tumor-derived Laminin-1 | |
| 25 μg | |
| Primary | |
| Detects the Hep-1 heparin binding domain of mouse Laminin A/alpha 1 subunit (1). | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| AL-4 | |
| Western Blot, Immunohistochemistry 8-25 ug/mL | |
| LAMA, LAMA1, LAMA1 laminin, alpha 1, S-LAM-alpha | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 284217 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction