missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Kirrel2/NEPH3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2930-SP
This item is not returnable.
View return policy
Description
Kirrel2/NEPH3 Polyclonal specifically detects Kirrel2/NEPH3 in Mouse samples. It is validated for Western Blot.
Specifications
| Kirrel2/NEPH3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp564A1164, Filtrin, FILTRINkin of IRRE-like protein 2, kin of IRRE like 2 (Drosophila), Kin of irregular chiasm-like protein 2, MGC15718, NEPH3, NEPH3Nephrin-like protein 3, nephrin-like 3, nephrin-like gene 1, NLG1, NLG1kin of irregular chiasm-like 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 84063 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q7TSU7 | |
| KIRREL2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Kirrel2/NEPH3 Ser19-Asp501 Accession # Q7TSU7 | |
| 25 μg | |
| Primary | |
| Detects mouse Kirrel2/NEPH3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human Kirrel2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction