missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Kell Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1454
This item is not returnable.
View return policy
Description
Kell Polyclonal specifically detects Kell in Mouse samples. It is validated for Western Blot.
Specifications
| Kell | |
| Polyclonal | |
| Unconjugated | |
| Q9EQF2 | |
| KEL | |
| Mouse myeloma cell line NS0-derived recombinant mouse Kell Phe50-Trp713 Accession # Q9EQF2 | |
| 100 μg | |
| Primary | |
| Detects mouse Kell in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CD238, CD238 antigen, EC 3.4.24.-, ECE3, KEL, Kell blood group, kell blood group antigen, kell blood group glycoprotein, Kell blood group, metalloendopeptidase, Kell blood group, metallo-endopeptidase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3792 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction