missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Kell Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1454-SP
This item is not returnable.
View return policy
Description
Kell Monoclonal specifically detects Kell in Mouse samples. It is validated for Western Blot.
Specifications
| Kell | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9EQF2 | |
| KEL | |
| Mouse myeloma cell line NS0-derived recombinant mouse Kell Phe50-Trp713 Accession # Q9EQF2 | |
| 25 μg | |
| Primary | |
| Detects mouse Kell in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ECE-2, rhNeprilysin, or recombinant mouse Neprilysin is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 214711 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD238, CD238 antigen, EC 3.4.24.-, ECE3, KEL, Kell blood group, kell blood group antigen, kell blood group glycoprotein, Kell blood group, metalloendopeptidase, Kell blood group, metallo-endopeptidase | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3792 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction