Learn More
Mouse Kell Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand:  R&D Systems MAB1454
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18119150
  • £442.75 / 500µg
Estimated Shipment: 07-06-2024
to see stock.
Description
Description
Kell Monoclonal specifically detects Kell in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Kell
Monoclonal
LYOPH
Western Blot 1 ug/mL
Q9EQF2
KEL
Mouse myeloma cell line NS0-derived recombinant mouse Kell Phe50-Trp713 Accession # Q9EQF2
500 μg
Primary
Detects mouse Kell in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ECE-2, rhNeprilysin, or recombinant mouse Neprilysin is observed.
Mouse
Purified
Western Blot
214711
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD238, CD238 antigen, EC 3.4.24.-, ECE3, KEL, Kell blood group, kell blood group antigen, kell blood group glycoprotein, Kell blood group, metalloendopeptidase, Kell blood group, metallo-endopeptidase
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
3792
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a