missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse JMJD6/PSR Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1994
This item is not returnable.
View return policy
Description
JMJD6/PSR Polyclonal specifically detects JMJD6/PSR in Mouse samples. It is validated for Western Blot.
Specifications
| JMJD6/PSR | |
| Polyclonal | |
| Unconjugated | |
| Q9ERI5 | |
| JMJD6 | |
| Mouse myeloma cell line NS0-derived recombinant mouse JMJD6/PSR Ala289-Arg403 Accession # Q9ERI5 | |
| 100 μg | |
| Primary | |
| Detects mouse JMJD6/PSR in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| bifunctional arginine demethylase and lysyl-hydroxylase JMJD6, EC 1.14.11, EC 1.14.11.-, Histone arginine demethylase JMJD6, JmjC domain-containing protein 6, jumonji domain containing 6, Jumonji domain-containing protein 6, KIAA0585PSR, Lysyl-hydroxylase JMJD6, Peptide-lysine 5-dioxygenase JMJD6, Phosphatidylserine receptorPTDSR, Protein PTDSR, PSR, PTDSR, PTDSR1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23210 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction