missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse JAM-C Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF1213
This item is not returnable.
View return policy
Description
JAM-C Polyclonal specifically detects JAM-C in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction, Immunocytochemistry.
Specifications
| JAM-C | |
| Polyclonal | |
| Unconjugated | |
| Q9D8B7 | |
| JAM3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse JAM-C Val32-Asn241 Accession # Q9D8B7 | |
| 100 μg | |
| Primary | |
| Detects mouse JAM-C in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human JAM-C and less than 2% cross-reactivity with recombinant mouse (rm) JAM-A and rmJAM-B is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction, Immunocytochemistry 5-15 ug/mL | |
| CD323, JAM-2, JAM3, JAMC, JAM-CFLJ14529, junctional adhesion molecule 3JAM-3, junctional adhesion molecule C | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 83700 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction