missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse JAM-B/VE-JAM Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
£167.00 - £1510.00
Specifications
| Antigen | JAM-B/VE-JAM |
|---|---|
| Clone | 150005 |
| Dilution | Neutralization 0.1-0.5 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18734289
![]() |
R&D Systems
MAB9881-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18729128
![]() |
R&D Systems
MAB9881-100 |
100 μg |
£380.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18711537
![]() |
R&D Systems
MAB9881-500 |
500 μg |
£1510.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JAM-B/VE-JAM Monoclonal specifically detects JAM-B/VE-JAM in Mouse samples. It is validated for Neutralization.Specifications
| JAM-B/VE-JAM | |
| Neutralization 0.1-0.5 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q9JI59 | |
| 58494 | |
| Mouse myeloma cell line NS0-derived recombinant mouse JAM-B/VE-JAM Phe29-Asn236 (predicted) Accession # Q9JI59 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse JAM-B/VE-JAM in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human JAM-1, -2, or -3 is observed. |
| 150005 | |
| Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C21orf43, CD322, CD322 antigen, JAM2, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAM, VE-JAMVEJAMCD322 | |
| JAM2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title