missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse JAM-A Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF1077-SP
This item is not returnable.
View return policy
Description
JAM-A Polyclonal specifically detects JAM-A in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| JAM-A | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD321, CD321 antigen, F11 receptor, F11R, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1, PAM-1KAT, Platelet adhesion molecule 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 50848 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| O88792 | |
| F11R | |
| Mouse myeloma cell line NS0-derived recombinant mouse JAM-A Lys27-Ala242 Accession # O88792 | |
| 25 μg | |
| Primary | |
| Detects mouse JAM-A in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant human JAM-A, recombinant mouse (rm) JAM-B, and rmJAM-C is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction