missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin alpha E/CD103 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1990
This item is not returnable.
View return policy
Description
Integrin alpha E/CD103 Polyclonal specifically detects Integrin alpha E/CD103 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Integrin alpha E/CD103 | |
| Polyclonal | |
| Unconjugated | |
| Q60677 | |
| ITGAE | |
| Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha E/CD103 Phe20-Arg1113 Accession # Q60677 | |
| 100 μg | |
| Primary | |
| Detects mouse Integrin alpha E/CD103 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), ITGAE, MGC141996, Mucosal lymphocyte 1 antigen | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3682 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction