missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin alpha 5/CD49e Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1754-SP
This item is not returnable.
View return policy
Description
Integrin alpha 5/CD49e Polyclonal specifically detects Integrin alpha 5/CD49e in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Integrin alpha 5/CD49e | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5, very late activation protein 5, alpha subunit, VLA-5, VLA-5 alpha | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3678 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P11688 | |
| ITGA5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha 5/CD49e Phe45-Asn (Gln68Glu) Accession # P11688 | |
| 25 μg | |
| Primary | |
| Detects mouse Integrin alpha 5/CD49e in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) Integrin alpha 5 is observed, 15% cross-reactivity with rhIntegrin alpha 2 and rhIntegrin alpha M is observed, and 5% cross-reactivity with rhIntegrin alpha E is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction