missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin alpha 11 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6498-SP
This item is not returnable.
View return policy
Description
Integrin alpha 11 Polyclonal specifically detects Integrin alpha 11 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Integrin alpha 11 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HsT18964, integrin alpha-11, integrin, alpha 11, ITGA11 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 22801 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P61622 | |
| ITGA11 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 11 Phe19-Pro1141 Accession # P61622 | |
| 25 μg | |
| Primary | |
| Detects mouse Integrin alpha 11 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) Integrin alpha 11 is observed, and less than 2% cross-reactivity with recombinant mouse (rm) Integrin alph | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction