missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse iNOS ELISA Kit (Colorimetric)

Product Code. 18270107 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18270107 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18270107 Supplier Novus Biologicals™ Supplier No. NBP280256

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This kit recognizes Mouse

No significant cross-reactivity or interference between Mouse NOS2/iNOS and analogues was observed.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29477, P29477
Assay Range 0.313 to 20 ng/mL
Assay Sensitivity 0.188 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 4843
Gene Symbol NOS2
Host Species Mouse
Quantity 1 Kit
Research Discipline Adaptive Immunity, Apoptosis, Cancer, HIF Target Genes, Hypoxia, Inflammation, Lipid and Metabolism, Neuroscience, Neurotransmission
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2
Target iNOS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.