missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse IL-7R alpha/CD127 (soluble) Fc Recombinant Protein, PeproTech®
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
Description
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
Specifications
Specifications
| Accession Number | P10168, P16872 |
| For Use With (Application) | ELISA |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 16196, 16197 |
| Molecular Weight (g/mol) | 51 kDa |
| Name | IL-7 |
| Quantity | 100 μg |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction