missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-7R alpha/CD127 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems FAB7473R-100UG
This item is not returnable.
View return policy
Description
IL-7R alpha/CD127 Monoclonal antibody specifically detects IL-7R alpha/CD127 in Mouse samples. It is validated for Flow Cytometry
Specifications
| IL-7R alpha/CD127 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
| NS0 mouse myeloma cell line transfected with mouse IL-7 R alpha /CD127, Glu21-Asp239, Accession # P16872 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 1140A | |
| Flow Cytometry 5 uL/10^6 cells | |
| P16872 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3575 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction