missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-7, Recombinant Protein, eBioscience™
Recombinant Protein
Brand: Affymetrix eBioscience 14-8071-62
Code : NEW
Additional Details : Weight : 0.09500kg
Description
The protein encoded by this gene is a cytokine important for B and T cell development. This cytokine and the hepatocyte growth factor (HGF) form a heterodimer that functions as a pre-pro-B cell growth-stimulating factor. This cytokine is found to be a cofactor for V(D)J rearrangement of the T cell receptor beta (TCRB) during early T cell development. This cytokine can be produced locally by intestinal epithelial and epithelial goblet cells, and may serve as a regulatory factor for intestinal mucosal lymphocytes. Knockout studies in mice suggested that this cytokine plays an essential role in lymphoid cell survival. Description: Mouse IL-7 (also called Lymphopoietin 1, LP-1), a pre-B cell growth factor, is an ∽25kDa factor produced by bone marrow and thymic stromal cells, and spleen cells. IL-7 is a stromal cell-derived growth factor for progenitor B cells and T cells. The main population in the thymus responsive to IL-7 is CD4^-CD8^-. It also stimulates proliferation and differentiation of mature T cells and NK cells. Applications Reported: Recombinant mouse IL-7 is biologically active. Applications Tested: The ED50 of this protein, as measured by 2E8 cell proliferation assay, is less than or equal to 1.6ng/mL. This corresponds to a specific activity of greater than or equal to 6.3 x 10e5 Units/mg. Source: E. coli expressed amino acids Glu 26-Ile 154 of mature mouse IL-7 accession # NM_008371. Bioactivity: The ED50 of this protein, as measured by 2E8 cell proliferation assay, is less than or equal to 1.6ng/mL. This corresponds to a specific activity of greater than or equal to 6.3 x 10e5 Units/mg. Endotoxin: Less than 0.01ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE. Molecular Weight: The methionylated polypeptide has a predicted molecular mass of 15,028. The DTT reduced protein migrates as a 15kDa polypeptide. The non-reduced protein has slightly higher mobility on non-reducing SDS-PAGE. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2μm post-manufacturing filtered.Specifications
P10168 | |
Cytokine Bioassay, Control, Functional Assay | |
16196 | |
Mouse IL-7 | |
SDS-PAGE | |
E. coli | |
RUO | |
Less than 0.01ng/μg cytokine as determined by the LAL assay. | |
Il7 | |
Mouse | |
Ms | |
Recombinant | |
Recombinant |
0.1mg/mL | |
protein with 0.1M glycine and no preservative; pH 3.0 | |
The methionylated polypeptide has a predicted molecular mass of 15,028. The DTT reduced protein migrates as a 15kDa polypeptide. The non-reduced protein has slightly higher mobility on non-reducing SDS-PAGE. | |
3.0 | |
5μg | |
-80°C | |
E. coli expressed amino acids glu 26-ile 154 of mature mouse IL-7 accession # NM_008371 | |
Mouse IL-7 | |
The ED50 of this protein, as measured by 2E8 cell proliferation assay, is less than or equal to 1.6ng/mL. This corresponds to a specific activity of greater than or equal to 6.3 x 105 Units/mg. | |
Unconjugated | |
E. coli | |
Liquid | |
Affinity chromatography |
For Research Use Only.