missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-6R alpha Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems
Additional Details : Weight : 0.00970kg
Description
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
IL-6R alpha | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3570 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Flow Cytometry, Immunocytochemistry, CyTOF | |
255821 | |
Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
P22272 | |
Il6ra | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-6 R alpha Leu20-Glu357 Accession # P22272 | |
500 μg | |
Primary | |
Detects mouse IL-6 R alpha in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human IL-6 R alpha is observed. | |
Mouse | |
Purified |
For Research Use Only