missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-6 Quantikine ELISA Kit
Mouse IL-6 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
£534.54 - £1852.00
Specifications
Assay Sensitivity | 1.8 pg/mL |
---|---|
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | Solid Phase Sandwich ELISA |
Specificity | Natural and Recombinant Mouse IL-6 |
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
18792034
|
R&D Systems™
SM6000B |
1 Pk. (6 Plates) | Sufficient materials to run ELISAs on at least six 96-well plates |
£1852.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18702244
|
R&D Systems™
M6000B-1 |
1 Kit (1 Plate) | Sufficient materials to run ELISAs on at least six 96-well plates |
£534.54
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Mouse IL-6 Quantikine ELISA Kit quantifies mouse IL-6 in cell culture supernates (50 ul), serum (50 ul), heparin plasma (50 ul).Specifications
1.8 pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
Natural and Recombinant Mouse IL-6 | |
ELISA | |
Il6 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
IL-6 |
HRP | |
Solid Phase Sandwich ELISA | |
Mouse | |
3569 | |
Cell Culture Supernates (50uL),Serum (50uL),Heparin Plasma (50uL) | |
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2 A |