missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Mouse IL-6 ELISA Kit

Product Code. 16801606 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Mouse
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Species unitSize
16801606 Mouse 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16801606 Supplier Proteintech™ Supplier No. KE1000796T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KE10007 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The Mouse IL6 ELISA kit is to be used to detect and quantify protein levels of endogenous mouse IL6.

  • Interleukin-6 (Il-6) is an interleukin that acts as both a pro-inflammatory and anti-inflammatory cytokine. Il-6 protein is secreted by a variety of cell types including T cells and macrophages as phosphorylated and variably glycosylated molecule.
  • Il-6 Plays an essential role in the final differentiation of B-cells into Ig-secreting cells involved in lymphocyte and monocyte differentiation.
  • It induces myeloma and plasmacytoma growth and induces nerve cells differentiation Acts on B-cells, T-cells, hepatocytes, hematopoietic progenitor cells and cells of the CNS.
  • Il-6 is also considered a myokine, a cytokine produced from muscle, and is elevated in response to muscle contraction.
  • Il-6 has been shown to interact with interleukin-6 receptor and glycoprotein 130. Additionally, Il-6 is involved in hematopoiesis, bone metabolism, and cancer progression, and has been defined an essential role in directing transition from innate to acquired immunity.
  • The assay recognizes mouse IL6. An antibody specific for mouse IL6 has been pre-coated onto the microwells.
  • The mouse IL6 protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for mouse IL6 is added to detect the captured mouse IL6 protein.
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent.
  • Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450nm with the correction wavelength set at 630 nm.

Specifications

Accession Number NP_112445
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 3.8 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Mouse
Gene ID (Entrez) 16193
Host Species Mouse
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID P08505
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96 TEST Mouse IL-6 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Tetramethylbenzidine, sulphuric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.