missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-4R alpha Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB530
This item is not returnable.
View return policy
Description
IL-4R alpha Monoclonal specifically detects IL-4R alpha in Mouse samples. It is validated for Western Blot.
Specifications
| IL-4R alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD124, CD124 antigen, IL-4 R alpha, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3566 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 129801 | |
| Western Blot 1 ug/mL | |
| P16382 | |
| Il4ra | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-4 R alpha Ile26-Arg233 Accession # P16382 | |
| 500 μg | |
| Primary | |
| Detects mouse IL-4 R alpha in direct ELISAs and Western blots. Shows less than 1% cross-reactivity with recombinant human IL-4 R alpha . | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction