missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-38/IL-1F10 Antibody, R&D Systems™

Rat Monoclonal Antibody
£188.00 - £444.00
Specifications
| Antigen | IL-38/IL-1F10 |
|---|---|
| Clone | 798038R |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18754607
![]() |
R&D Systems
MAB77742-SP |
25 μg |
£188.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18793837
![]() |
R&D Systems
MAB77742-100 |
100 μg |
£444.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for ELISA.Specifications
| IL-38/IL-1F10 | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| Q8R459 | |
| 84639.0 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C, as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 6 months, -20 to -70 °C under sterile conditions after opening. |
| 798038R | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 | |
| E. coli-derived recombinant mouse IL-38/IL-1F10, Met1-Arg152, Accession No. Q8R459 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-38/IL-1F10 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title