missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-38/IL-1F10 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB77741S-100UG
This item is not returnable.
View return policy
Description
IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IL-38/IL-1F10 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 | |
| E.coli-derived recombinant mouse IL-38/IL-1F10 Met1-Arg152 Accession # Q8R459 | |
| RUO | |
| 84639 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 798036R | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8R459 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse IL-38/IL-1F10 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction