missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-38/IL-1F10 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB7774U-100UG
This item is not returnable.
View return policy
Description
IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-38/IL-1F10 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8R459 | |
| IL1F10 | |
| E. coli-derived recombinant mouse IL-38/IL-1F10 Met1-Arg152 Accession # Q8R459 | |
| 100 μg | |
| Primary | |
| Detects mouse IL-38/IL-1F10 in ELISA and Western Blot. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 798036 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84639 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction