missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human Brain-Derived Neurotrophic Factor (BDNF), Recombinant Protein
Click to view available options
Quantity:
5μg
Unit Size:
5µg
Beskrivning
Brain-Derived Neurotrophic Factor (BDNF) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, proBDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation.
Specifikationer
Specifikationer
| Accession Number | NM_001709 |
| Concentration | 0.1mg/mL |
| For Use With (Application) | Cytokine Bioassay |
| Formulation | Sterile liquid; 20mM NaCitrate, 300mM NaCl, 1% BSA, pH 4.0, 0.22μm filtered |
| Molecular Weight (g/mol) | 15.4kDa |
| Name | Brain-Derived Neurotrophic Factor (BDNF) |
| Purification Method | SDS-PAGE |
| Quantity | 5μg |
| Source | E. coli |
| Storage Requirements | Store at less than or equal to -70°C. |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering