missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-3 Recombinant Protein

Recombinant Mouse Interleukin 3 (IL3) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC0035

Code : 50

Additional Details : Weight : 0.02100kg

 View more versions of this product

Product Code. 10670154

  • £366.00 / Each
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Mouse IL-3 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IL3 provides the cytokine connection between the immune system and the hematopoietic system. For example, IL3 supports the proliferation and development of almost all types of hematopoietic progenitor cells and can act as a chemoattractant for eosinophils.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of NFS-60 cells

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture
16.1 kDa
25μg
Mouse
Carrier-Free
25 μg
E. coli
Recombinant, Ligand
Interleukins
Gibco™
16187
Sequential Chromatography
IL-3
Store in refrigerator (2–8°C).
< 0.1 ng/μg
Wet Ice
Cytokines & Receptors
Lyophilized
Immunology
95 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.