missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-28A Mouse ELISA Kit
For the quantitative detection of mouse IL-28.
Brand: Invitrogen™ BMS6028
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-28
Biotin-Conjugate anti-mouse IL-28 monoclonal antibody
Streptavidin-HRP
Mouse IL-28 Standard lyophilized, 4000pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x
Description
IL-28A belongs to the IFN lambda family, a novel family of cytokines within the IL-10 superfamily. The three members of this family are IL-29 (IFN lambda 1), IL-28A (IFN lambda 2), and IL-28B (IFN lambda 3), and are also known as the type III Interferons.The IFN lambdas signal through a heterodimeric receptor of which one subunit, IL-10R2, is shared with other members of the superfamily. The second subunit, IFN lambda R1 or IL-28R alpha, is unique to the IFN lambdas. Signaling occurs through the Jak/STAT pathway in a similar manner as the type I IFN (IFN alpha/beta) and activates many of the same genes despite low sequence homology between the cytokines and receptors in the two families. Both IFN families display antiviral activity through the induction of antiviral protein production in target cells and the upregulation of MHC class I expression. These proteins also exhibit antiproliferative and antitumor effects, making them a possible alternative to IFN alpha cancer therapies. Unlike the type I IFN, which are able to stimulate most cells, response to IFN lambda stimulation appears to be limited to dendritic and some tumor cells due to the limited expression of IFN lambda R1. Another notable difference is the ability of the IFN lambda stimulation to drive dendritic cells towards the production of CD4+CD25+FoxP3+ regulatory T cells, suggesting a possible immunoregulatory role.Specifications
Q4VK74 | |
Biotin | |
330496 | |
ELISA Kit | |
12.85 pg/mL | |
Plasma, 50 μL|Serum, 50 μL|Supernatant, 50 μL | |
1 hr. 20 min. | |
Mouse | |
IFNL2, IFNL3 Interleukin 28, IL-28A, IL-28B | |
5.6% | |
Mouse | |
HRP |
31.3-2,000 pg/mL | |
ELISA | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
50 μL | |
2°C to 8°C | |
3 hr. 30 min. | |
EG330496,IL-28A,Il28a | |
7.2% | |
96 Tests | |
Supernatant | |
Colorimetric Microplate Reader |
For Research Use Only