missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-27 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC18342R-100UG
This item is not returnable.
View return policy
Description
IL-27 Monoclonal specifically detects IL-27 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-27 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O35228 (EBI3) & Q8K3I6 (p28) | |
| IL27 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-27 heterodimer Tyr19-Pro228 (EBI3), Phe29-Ser234 (p28) Accession # O35228 (EBI3) & Q8K3I6 (p28) | |
| RUO | |
| 246778 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 355025 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| IL27, IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse IL-27 in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse (rm) IL-27 p28, rmIL-27 EBI3, recombinant human (rh) IL-27 p28, and rhIL-27 EBI3 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction