missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-23 p19 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
£168.00 - £487.00
Specifications
| Antigen | IL-23A/IL-23 P19 |
|---|---|
| Clone | 320229 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18705528
![]() |
R&D Systems
MAB1619-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18132471
|
R&D Systems
MAB1619 |
500 μg |
£487.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
IL-23A/IL-23 P19 Monoclonal specifically detects IL-23A/IL-23 P19 in Mouse samples. It is validated for Western Blot.Specifikationer
| IL-23A/IL-23 P19 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL23, IL-23, IL-23 p19, IL-23 p19/IL-12 p40, IL23A, IL-23A, IL-23-A, IL-23p19, IL-23p19/IL-12p40, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRF, SGRFIL-23 subunit alpha | |
| IL23A | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 320229 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P43432 (p40) & Q9EQ14 (p19) | |
| 51561 | |
| E. coli-derived recombinant mouse IL-23 p19 and S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-23 heterodimers Met23-Ser335 (p40) & Leu20-Ala196 (p19) Accession # P43432 (p40) & Q9EQ14 (p19) | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-23 p19/p40 heterodimers and p19 subunits in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human p19 is observed. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel