missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-23 DuoSet ELISA
Click to view available options
Quantity:
5 Plates
Unit Size:
Pack of 5
Description
Includes
Mouse IL-23 Capture Antibody; Mouse IL-23 Detection Antibody; Mouse IL-23 Standard; Streptavidin-HRP
The Mouse IL-23 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IL-23A/IL-23 P19.
Specifications
Specifications
| Quantity | 5 Plates |
| Species | Mouse |
| Conjugate | Biotin |
| For Use With (Application) | ELISA |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction