missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-22 Antibody, R&D Systems™

Rat Monoclonal Antibody
£168.00 - £409.00
Specifications
| Antigen | IL-22 |
|---|---|
| Clone | 924324 |
| Dilution | Neutralization 0.5-6 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18765518
![]() |
R&D Systems
MAB5821-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18718049
![]() |
R&D Systems
MAB5821 |
100 μg |
£409.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-22 Monoclonal specifically detects IL-22 in Mouse samples. It is validated for Neutralization.Specifications
| IL-22 | |
| Neutralization 0.5-6 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| IL22 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 924324 | |
| Neutralization | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9JJY9 | |
| 50616 | |
| E. coli-derived recombinant mouse IL-22 Leu34-Val179 Accession # Q9JJY9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-22 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title