missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-21 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB594-SP
This item is not returnable.
View return policy
Description
IL-21 Monoclonal specifically detects IL-21 in Mouse samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| IL-21 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 59067 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Flow Cytometry, CyTOF | |
| 149204 | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9ES17 | |
| IL21 | |
| E. coli-derived recombinant mouse IL-21 Pro25-Ser146 Accession # Q9ES17 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-21 in Western blots. Does not cross-react with recombinant human IL-21, recombinant mouse (rm) IL-15, or rmIL-12. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction