missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-20R alpha Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF1736
Additional Details : Weight : 0.00970kg
Description
IL-20R alpha Polyclonal specifically detects IL-20R alpha in Mouse samples. It is validated for Western Blot.Specifications
IL-20R alpha | |
Polyclonal | |
Unconjugated | |
Q6PHB0 | |
Il22ra2 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-20 R alpha Val33-Lys253 Accession # Q6PHB0 | |
100 μg | |
Primary | |
Detects mouse IL-20 R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) IL-20 R alpha is observed and less than 5% cross-reactivity with rhIL-20 R beta , rhIFN-gamma RI, rhIFN-gamma RII, and rhIL-10 R beta is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20 R alpha, IL20R alpha, IL-20R1, IL-20R1IL-20 receptor subunit alpha, IL20RA, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7 | |
Sheep | |
Affinity Purified | |
RUO | |
53832 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only