missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-2 R beta Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB589
This item is not returnable.
View return policy
Description
IL-2 R beta Monoclonal specifically detects IL-2 R beta in Mouse samples. It is validated for Western Blot.
Specifications
| IL-2 R beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-15 R beta, IL-2 receptor subunit beta, IL2R beta, IL-2R subunit beta, IL2RB, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3560 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 130128 | |
| Western Blot 1 ug/mL | |
| P16297 | |
| IL2RB | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-2 R beta Ala27-Glu240 Accession # P16297 | |
| 500 μg | |
| Primary | |
| Detects mouse IL-2 R beta in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2 R beta , rhIL-2 R alpha , recombinant mouse (rm) IL-2 R gamma , rmIL-21 R, or rhIL-15 R is observed. In Western blots under reducing conditions, approximately 50% cross-reactivity with rhIL-2 R beta is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction