missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-2 ELISA Kits

Product Code. 10300974
Change view
Click to view available options
Quantity:
1 Plate Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10300974 1 Plate Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10300974 Supplier Invitrogen™ Supplier No. EMIL2

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Measure levels of mouse interleukin-2 in mouse serum and cell culture supernatant samples using 96-well plates.

Assay IL-2 (mouse interleukin-2) levels in mouse serum and culture supernatant samples with our Thermo Scientific™ Mouse IL-2 ELISA Kits.

These mouse IL-2 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-2 (IL-2) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. The antibody-pair kit consists of the matched IL-2 antibody pairs and IL-2 standard only. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant mouse interleukin-2 (IL-2)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 34 to 850pg/mL

  • Sensitivity: < 3pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum

  • Total Assay Time: 3.5 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with mouse IL-3, IL-4,IL-5, IL-6, IL-10, IFNγ, GM-CSF, TNFα or human IL-2

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 34 to 850pg/mL
Assay Sensitivity <3pg/mL
Description Mouse IL-2 ELISA Kit
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Sample Type Cell Culture Supernatant, Serum
Specificity No significant cross-reaction with mouse IL-3, IL-4,IL-5, IL-6, IL-10, IFNg, GM-CSF, TNFa or human IL-2
Sufficient For 96 microplate wells
Kit Contents TMB Substrate Solution, 13mL, Stop Solution, 13mL, Anti-Mouse IL-2 Precoated 96-well Strip Plate, 1 ea, Standard Diluent, 12mL, Plate Reagent, 12mL, Conjugate Dilution Buffer, 14mL, Anti-IL2-HRP Conjugate Concentrate, 0.10mL, Adhesive Plate Covers, 6 ea, 30X Wash Buffer, 50mL, Lyophilized Recombinant Mouse IL-2 Standard, 2 vials
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Quantity 1 Plate Kit
Sample Volume 50 μL
Target Natural and recombinant mouse interleukin-2 (IL-2)
Total Assay Time 3.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.