missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-1Rrp2/IL-1R6 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2354-SP
This item is not returnable.
View return policy
Description
IL-1Rrp2/IL-1R6 Polyclonal specifically detects IL-1Rrp2/IL-1R6 in Mouse samples. It is validated for Western Blot.
Specifications
| IL-1Rrp2/IL-1R6 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-1 R6, IL-1 Rrp2, IL1RL2, IL-1RL2, IL1Rrp2, IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8808 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q9ERS7 | |
| Il1rl2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-1 Rrp2/IL-1 R6 Asp22-Arg338 (Ala158Val) Accession # Q9ERS7 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-1 Rrp2/IL-1 R6 in direct ELISAs and Western blots. In direct ELISAs, less than 60% with recombinant rat IL-1 Rrp2 and recombinant human IL-1 Rrp2 is observed and less than 5% cross-reactivity with recombinant mouse (rm) IL-1 R4 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction