missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-18R alpha/IL-1R5 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1216T-100UG
This item is not returnable.
View return policy
Description
IL-18R alpha/IL-1 R5 Monoclonal specifically detects IL-18R alpha/IL-1 R5 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IL-18R alpha/IL-1 R5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q61098 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse IL-18 R alpha/IL-1 R5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) IL-1 RI, rmIL-1 RII, recombinant human (rh) IL-1 R3, rhIL-1 R4, recombinant rat (rr) IL-1 R6, rmIL-1 R7, rhIL-1 R8, rhIL-1 R9, or rmSIGIRR is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 112614 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Il18rap | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha/IL-1 R5 Ser19-Gly326 Accession # Q61098 | |
| RUO | |
| 8809 | |
| Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction