missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-18 R alpha/IL-1 R5 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF856
This item is not returnable.
View return policy
Description
IL-18R alpha/IL-1 R5 Polyclonal specifically detects IL-18R alpha/IL-1 R5 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.
Specifications
| IL-18R alpha/IL-1 R5 | |
| Polyclonal | |
| Unconjugated | |
| Q61098 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 8809 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Neutralization, CyTOF | |
| LYOPH | |
| Western Blot 0.1 μg/mL, Flow Cytometry 0.25 μg/106 cells, Neutralization 0.2 to 0.8 μg/mL, CyTOF-ready | |
| Il18rap | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha/IL-1 R5 Ser19-Gly326 Accession # Q61098 | |
| 100 μg | |
| Primary | |
| Detects mouse IL-18 R alpha in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human IL-18 R is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction