missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-17RA/IL-17R Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF448-SP
This item is not returnable.
View return policy
Description
IL-17RA/IL-17R Polyclonal specifically detects IL-17RA/IL-17R in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| IL-17RA/IL-17R | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL17RA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-17 RA/IL-17 R Extracellular domain | |
| 25 μg | |
| Primary | |
| Detects mouse IL-17 RA/IL-17 R in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human IL-17 RA/IL-17 R is observed and less than 1% cross-reactivity with recombinant mouse (rm) IL-17 RC and rmIL-17 RD is observed. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23765 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction