missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-17F Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF2057
This item is not returnable.
View return policy
Description
IL-17F Polyclonal specifically detects IL-17F in Mouse samples. It is validated for Western Blot, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-17F | |
| Polyclonal | |
| Unconjugated | |
| NP_665855 | |
| IL17F | |
| E. coli-derived recombinant mouse IL-17F Arg29-Ala153 Accession # NP_665855 | |
| 100 μg | |
| Primary | |
| Detects mouse IL-17F in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human IL-17F, recombinant mouse (rm) IL-17, rmIL-17B, rmIL-17C, rmIL-17D, and rmIL-17E is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Cytokine ML-1, IL17F, IL-17Finterleukin-17F, interleukin 17F | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 112744 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction