missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-25/IL-17E Mouse ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ BMS6046TEN

Code : NEW

Additional Details : Weight : 0.47500kg

 View more versions of this product

Product Code. 15592007

  • £2380.00 / Pack of 10


Estimated Shipment
08-10-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-17E (IL-25)
Biotin-Conjugate anti-mouse IL-17E (IL-25) monoclonal antibody
Streptavidin-HRP
Mouse IL-17E (IL-25) Standard lyophilized, 500pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

ELISA
50 μL
Interleukin-17E, Interleukin-25
Mouse
0.9pg/mL
IL-17E (IL-25)
10 x 96 Tests
Serum
Videos
Documents