missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-17D Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB22741-100
This item is not returnable.
View return policy
Description
IL-17D Monoclonal antibody specifically detects IL-17D in Mouse samples. It is validated for ELISA
Specifications
| IL-17D | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| FLJ30846, IL17D, IL-17Dinterleukin 27, IL-22, IL-27interleukin-17D, IL27interleukin-27, interleukin 17D, Interleukin-27 | |
| E. coli-derived recombinant mouse IL-17D, Ala25-Arg205, Accession # NP_665836 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 312708 | |
| ELISA | |
| NP_665836 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 53342 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction