missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-17A/F Heterodimer DuoSet ELISA
Click to view available options
Quantity:
5 Plates
Unit Size:
1 set
Description
The Mouse IL-17A/F Heterodimer DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IL-17A/F Heterodimer.
Specifications
Specifications
| Detectable Analytes | IL-17A/F Heterodimer |
| Assay | ELISA Development Kit |
| Conjugate | Biotin |
| Cross Reactivity | < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules. |
| Immunoassay Kit Format | 96-well microplate (sold separately) |
| For Use With (Application) | ELISA |
| Host Species | Mouse |
| Measuring Range | 46.9-3,000pg/mL |
| Quantity | 5 Plates |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction