missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-16 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1727
This item is not returnable.
View return policy
Description
IL-16 Polyclonal specifically detects IL-16 in Mouse samples. It is validated for Western Blot.
Specifications
| IL-16 | |
| Polyclonal | |
| Unconjugated | |
| O54824 | |
| IL16 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-16, Ser1205-Ser1322, Accession # O54824 | |
| 100 μg | |
| Primary | |
| Detects mouse IL-16 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human IL-16 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| FLJ16806, FLJ42735, FLJ44234, HsT19289, IL16, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCF, LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3603 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction