missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-12 R beta 2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
£171.00 - £366.00
Specifications
| Antigen | IL-12 R beta 2 |
|---|---|
| Clone | 918102 |
| Dilution | Blockade of Receptor-ligand Interaction 30-180 ng/mL |
| Applications | Blockade of Receptor-ligand Interaction |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18759597
![]() |
R&D Systems
MAB8650-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18722568
![]() |
R&D Systems
MAB8650 |
100 μg |
£366.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-12 R beta 2 Monoclonal specifically detects IL-12 R beta 2 in Mouse samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
| IL-12 R beta 2 | |
| Blockade of Receptor-ligand Interaction 30-180 ng/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P97378 | |
| 3595 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-12 R beta 2 Met1-Asn637 Accession # P97378 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-12 R beta 2 in direct ELISAs. |
| 918102 | |
| Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL12R beta 2, IL-12R subunit beta-2, IL12RB2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2 | |
| Il12rb2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title