missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-12 p80 Recombinant Protein, PeproTech®

Product Code. 17870003
Click to view available options
Quantity:
10 μg
10 x 50 μg
2 μg
2 x 50 μg
20 x 50 μg
5 x 50 μg
50 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17870003

Brand: Gibco™ 21012P80H50UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

210-12P80H-100UG will be provided as 2 x 50 μg (210-12P80H-50UG). 210-12P80H-1MG will be provided as 20 x 50 μg (210-12P80H-50UG). 210-12P80H-250UG will be provided as 5 x 50 μg (210-12P80H-50UG). 210-12P80H-500UG will be provided as 5 x 100 μg (210-12P80H-50UG). Recombinant Murine IL-12 p80 is an 80.0 kDa, disulfide linked homodimer of two p40 chains of IL-12 that consists of 626 amino acid residues and less than 5% of the IL-12 p40 monomer. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-12 is a disulfide-linked heterodimeric protein (p70), composed of two subunits, p35 and p40, which are encoded by two different genes. Accumulating data indicates that p40 secretion precedes that of IL-12 expression. In addition, to its ability to covalently bind to p35 to form IL-12, p40 can bind to p19 to form IL-23, or it can form the homodimer designated IL-12 p80. Elevated levels of IL-12 p80 correlate to macrophage recruitment and increased inflammation in asthma and respiratory viral infection models.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43432
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 16160
Molecular Weight (g/mol) 80 kDa
Name Mouse IL-12 p80
Quantity 50 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias IL-12; Interleukin
Common Name IL-12 p80
Gene Symbol IL12B
Biological Activity Determined by its ability to inhibit the proliferative effect of 2 ng/ml mIL-12 p70 in T cell-enriched PBMCs.
Conjugate Unconjugated
Recombinant Recombinant
Sequence p40 subunit: MWELEKDVYV VEVDWTPDAP GETVNLTCDT PEEDDITWTS DQRHGVIGSG KTLTITVKEF LDAGQYTCHK GGETLSHSHL LLHKKENGIW STEILKNFKN KTFLKCEAPN YSGRFTCSWL VQRNMDLKFN IKSSSSSPDS RAVTCGMASL SAEKVTLDQR DYEKYSVSCQ EDVTCPTAEE TLPIELALEA RQQNKYENYS TSFFIRDIIK PDPPKNLQMK PLKNSQVEVS WEYPDSWSTP HSYFSLKFFV RIQRKKEKMK ETEEGCNQKG AFLVEKTSTE VQCKGGNVCV QAQDRYYNSS CSKWACVPCR VRS
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.