missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-12 p70 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 14 publications
£178.00 - £1795.00
Specifications
| Antigen | IL-12 p70/IL-12A |
|---|---|
| Clone | 48110 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18795468
![]() |
R&D Systems
MAB419-SP |
25 μg |
£178.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713929
![]() |
R&D Systems
MAB419-100 |
100 μg |
£456.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18788059
![]() |
R&D Systems
MAB419-500 |
500 μg |
£1795.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-12 p70/IL-12A Monoclonal specifically detects IL-12 p70/IL-12A in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| IL-12 p70/IL-12A | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| CLMF p35, CLMF1, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL12, IL12 p70, IL-12 subunit p35, IL-12, subunit p35, IL12A, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
| IL12A | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 48110 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3592 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-12 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-12 in ELISAs and Western blots. Detects an epitope in the p35 subunit. In sandwich ELISAs, does not cross-react with recombinant mouse IL-12 p40. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title