missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-12 p40 Recombinant Protein, PeproTech®

Product Code. 17871623
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17871623

Brand: Gibco™ 21012P40H100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

210-12P40H-1MG will be provided as 2 x 500 μg (210-12P40H-500UG). Recombinant Murine IL-12 p40 is a 40.0 kDa protein consisting of 313 amino acid residues and less than 5% of the IL-12 p80 dimer. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL12B (IL-12 p40) encodes a subunit of interleukin 12, a cytokine that acts on T and natural killer cells, and has a broad array of biological activities. Interleukin 12 is a disulfide-linked heterodimer composed of the 40 kDa cytokine receptor like subunit encoded by this gene, and a 35 kDa subunit encoded by IL12A. This cytokine is expressed by activated macrophages that serve as an essential inducer of Th1 cells development. This cytokine has been found to be important for sustaining a sufficient number of memory/effector Th1 cells to mediate long-term protection to an intracellular pathogen. Overexpression of this gene was observed in the central nervous system of patients with multiple sclerosis (MS), suggesting a role of this cytokine in the pathogenesis of the disease. The promoter polymorphism of this gene has been reported to be associated with the severity of atopic and non-atopic asthma in children.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43432
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 16160
Molecular Weight (g/mol) 40 kDa
Name Mouse IL-12 p40
Quantity 100 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CLMF; CLMF p35; CLMF p40; CLMF2; cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; Il12; IL12 p40; IL-12 p40 subunit; IL-12 subunit p35; IL-12 subunit p40; IL12, subunit p40; IL-12.p40; IL12A; IL-12 A; Il12b; IL-12 B; Il12p40; IL-12p40; IL-12p40 subunit; ILN; IMD28; IMD29; Interleukin; Interleukin 12; interleukin 12 p40 subunit; interleukin 12 subunit p40; interleukin 12, p40; interleukin 12 B; interleukin 12 B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); Interleukin12; interleukin-12 beta chain; interleukin-12 beta chain precursor; interleukin-12 p40; interleukin-12 p40 chain; interleukin-12 p40 subunit; interleukin-12 subunit beta; interleukin-12 subunit p40; interleukin-12p40; interleukin-23 p40 subunit; natural killer cell stimulatory factor, 40 kD subunit; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; p40; RP23-388G23.1
Common Name IL-12 p40
Gene Symbol Il12b
Biological Activity Determined by its ability to inhibit the proliferative effect of 2 ng/ml mIL-12p70 in T cell-enriched PBMCs.
Conjugate Unconjugated
Recombinant Recombinant
Sequence p40 subunit: MWELEKDVYV VEVDWTPDAP GETVNLTCDT PEEDDITWTS DQRHGVIGSG KTLTITVKEF LDAGQYTCHK GGETLSHSHL LLHKKENGIW STEILKNFKN KTFLKCEAPN YSGRFTCSWL VQRNMDLKFN IKSSSSSPDS RAVTCGMASL SAEKVTLDQR DYEKYSVSCQ EDVTCPTAEE TLPIELALEA RQQNKYENYS TSFFIRDIIK PDPPKNLQMK PLKNSQVEVS WEYPDSWSTP HSYFSLKFFV RIQRKKEKMK ETEEGCNQKG AFLVEKTSTE VQCKGGNVCV QAQDRYYNSS CSKWACVPCR VRS
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.