missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-12 p40 Allele-specific Quantikine ELISA Kt

Product Code. 18772284
Click to view available options
Quantity:
2 Plates
6 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18772284

Brand: R&D Systems™ M1240

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse IL-12 p40 Allele-specific Quantikine ELISA Kt from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IL-12 p40 Allele-specific Quantikine ELISA Kt is a Solid Phase Sandwich ELISA that quantifies mouse IL-12/IL-23 p40 in cell culture supernates (50 ul), serum (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 11.70 - 750pg/mL
Assay Sensitivity 4pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and Recombinant Mouse IL-12/IL-23 p40
Sufficient For 2 x 96-well Plates
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 16160
Quantity 2 Plates
Sample Volume Cell Culture Supernates (50uL),Serum (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12 B, IL-12 BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12 B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.