missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-10 R beta Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5368-SP
This item is not returnable.
View return policy
Description
IL-10 R beta Polyclonal specifically detects IL-10 R beta in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| IL-10 R beta | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CDw210b, CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL10R beta, IL-10R2, IL-10R2IL-10R subunit beta, IL10RB, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3588 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q61190 | |
| IL10RB | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-10 R beta Ile21-Ser220 Accession # Q61190 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-10 R beta in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) IL-10 R beta is observed and less than 1% cross-reactivity with rhIL-10 R and recombinant mouse IL-10 R is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction